Läs mer
PF-06726304, MedChemExpress
Produktkod. 16483888
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
Förpackningsstorlek:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16483888 10 mg -
16430257 50 mg 50mg
16422896 25 mg -
16416617 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16483888 Leverantör Medchem Express Leverantörsnummer HY103682
MedChemExpress PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.
PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.
Specifikationer
Kemiskt namn eller material PF-06726304
CAS 1616287-82-1
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H21Cl2N3O3
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 15 mg/mL (33.61 mM)
LEDER O=C1N(CC2=C(C)C=C(C)NC2=O)CCC3=C1C(Cl)=C(C4=C(C)ON=C4C)C=C3Cl
Molekylvikt (g/mol) 446.33
Formel vikt 446.33
Procent renhet 99.64%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death