Läs mer
PF-3758309, MedChemExpress
Produktkod. 16491647
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16491647 50 mg -
16478187 5 mg 5mg
16426066 100 mg -
16425478 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16491647 Leverantör Medchem Express Leverantörsnummer HY13007
MedChemExpress PF-3758309 (PF-03758309) is a potent, orally available, and reversible ATP-competitive inhibitor of PAK4 (Kd= 2.7 nM; Ki=18.7 nM). PF-3758309 has the expected cellular functions of a PAK4 inhibitor: inhibition of anchorage-independent growth, induction of apoptosis, cytoskeletal remodeling, and inhibition of proliferation.
PF-3758309 (PF-03758309) is a potent, orally available, and reversible ATP-competitive inhibitor of PAK4 (Kd= 2.7 nM
Specifikationer
Kemiskt namn eller material PF-3758309
Fysisk form Solid
CAS 898044-15-0
Färg Off-White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C25H30N8OS
För användning med (applikation) Cancer-Kinase/protease
Synonym PF-03758309
Löslighetsinformation DMSO : ≥ 100 mg/mL (203.82 mM)
LEDER CC1=NC2=C(SC=C2)C(NC3=NNC4=C3CN(C(N[C@@H](C5=CC=CC=C5)CN(C)C)=O)C4(C)C)=N1
Molekylvikt (g/mol) 490.62
Formel vikt 490.62
Procent renhet 95.87%