Läs mer
Tocris Bioscience™ PF 431396
Beskrivning
Dual focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2) inhibitor (IC50 values are 2 and 11 nM respectively). Promotes osteoblast recruitment and activity, and stimulates bone formation in ovariectomized rats.
Specifikationer
Specifikationer
| Kemiskt namn eller material | PF 431396 |
| CAS | 717906-29-1 |
| Kvantitet | 10 mg |
| Molekylformel | C22H21F3N6O3S |
| Formel vikt | 506.5 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 506.50 |