Läs mer
PF-562271, MedChemExpress
Produktkod. 16486697
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16486697 5 mg
16453988 10 mg
16400347 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16486697 Leverantör Medchem Express Leverantörsnummer HY10459
MedChemExpress PF-562271 (VS-6062) is a potent, ATP-competitive and reversible FAK and Pyk2 kinase inhibitor with IC50s of 1.5 nM and 13 nM, respectively.
PF-562271 (VS-6062) is a potent, ATP-competitive and reversible FAK and Pyk2 kinase inhibitor with IC50s of 1.5 nM and 13 nM, respectively.
Specifikationer
Kemiskt namn eller material PF-562271
CAS 717907-75-0
Färg Pale Yellow
Anteckningar om renhetsgrad Research
Molekylformel C21H20F3N7O3S
Kvantitet 5 mg
Synonym VS-6062
Löslighetsinformation DMSO : ≥ 48 mg/mL (94.58 mM)
LEDER CS(=O)(N(C)C1=NC=CC=C1CNC2=NC(NC3=CC4=C(C=C3)NC(C4)=O)=NC=C2C(F)(F)F)=O
Molekylvikt (g/mol) 507.49
Formel vikt 507.49
Procent renhet 98.03%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease