missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16045-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.Specifications
PFDN4 | |
Polyclonal | |
Unconjugated | |
PFDN4 | |
C1, PFD4, PFDN4, prefoldin 4, prefoldin subunit 4, Protein C 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5203 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NQP4 | |
PFDN4 | |
PFDN4 Fusion Protein Ag8938 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |