Läs mer
Schmidlin™ PG Plus Rack Hydrogen Generator
Produktkod. 17768919
Encompass_Preferred
Change view
Flödeshastighet:
100 ml/min
250 ml/min
500 ml/min
Wattal:
65 W
115 W
180 W
Förpackningsstorlek:
Varje
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Flödeshastighet Wattal unitSize
17768919 100 ml/min 65 W Varje
17778919 250 ml/min 115 W Varje
17788919 500 ml/min 180 W Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17768919 Leverantör Schmidlin™ Leverantörsnummer DBRHG100EU
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
This generator produces a continuous supply of high purity hydrogen.
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Pressure: 11 barg (160 psig)
  • Unique permeation membrane drying system
  • USB connectivity
  • Easy to install with complete 'plug and play' functionality
  • Ideal for all GC detector applications
  • Meets and exceeds requirements for the most demanding GC applications
  • Superior hydrogen production with reliable long life cell
  • Minimal maintenance; no desiccant cartridges to change
  • PC monitoring for maintenance, diagnostics and remote connection
Specifikationer
Djup (metriskt) 500 cm
Flödeshastighet 100 ml/min
Höjd (metrisk) 13.3 cm
Max. Omgivningstemperatur 35°C
Utloppstryck 11 bar
Bredd (engelska) 19 in.
Bredd (metrisk) 48.5 cm
Anslutningar Outlet: 1/8 in.
Elektriska krav 110/120 V at 60 Hz, 220/240 V at 50 Hz
Kväve renhet >99.9996%
Wattal 65 W