Learn More
PGAM1 Mouse anti-Human, Unconjugated, Clone: 2H2A9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67470-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840116
  • 1236.20 SEK / 20µL
Estimated Shipment: 03-10-2022
to see stock.
Description
Description
The PGAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PGAM1. This antibody recognizes human, mouse, rat antigen. The PGAM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
PGAM1
Monoclonal
500 μg/mL
Mouse
Primary
Mouse, Rat, Human
Immunofluorescence, Immunohistochemistry, Western Blot
2H2A9
Unconjugated
20 μL
5223
IgG1