missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGCP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PGCP |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836264
|
Proteintech
16601-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826264
|
Proteintech
16601-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PGCP is a metallopeptidase that belongs to the peptidase M28 family. The encoded protein may catalyze the cleavage of dipeptides with unsubstituted terminals into amino acids.Specifications
PGCP | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q9WVJ3, Q9Y646 | |
10404, 54381 | |
PGCP Fusion Protein Ag9770 | |
Primary | |
-20°C | |
CPQ |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPQ, LCH1, Lysosomal dipeptidase, PGCP, plasma glutamate carboxypeptidase | |
Cpq | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |