Learn More
PGPEP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557245
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PESIDSIIDM DAVCKRVTTL GLDVSVTISQ DAGRYLCDFT YYTSLYQSHG RSAFVHVPPL GKPYNADQLG RALR Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Pyroglutamyl peptidase I (EC 3.4.19.3) catalyzes the hydrolysis of N-terminal pyroglutamyl residues from oligopeptides and proteins.Specifications
PGPEP1 | |
Polyclonal | |
Unconjugated | |
PGPEP1 | |
2810003H13Rik; 5-oxoprolyl-peptidase; Gm19458; PAPI; PAP-I; Pcp; PGI; pGlu-peptidase I; PGP; PGPEP1; PGPI; PGP-I; putative pyroglutamyl-peptidase I; pyroglutamyl aminopeptidase I; Pyroglutamyl-peptidase 1; pyroglutamyl-peptidase I; Pyrrolidone-carboxylate peptidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54858 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXJ5 | |
PGPEP1 | |
Recombinant protein corresponding to Human PGPEP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only