missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14327-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is expected to detect the endogenous PGT protein.
Prostaglandin transporter (PGT), also known as SLCO2A1, has been identified as a functional prostaglandin (PG) carrier that mediates either the release of newly-synthesized PGs by cells and/or the uptake of PGs prior to intracellular oxidation. Northern blot analysis revealed that PGT mRNA has broad tissue distribution, suggesting its role in the transport and/ or metabolic clearance of PG in diverse tissues (8787677).Specifications
PGT | |
Polyclonal | |
Unconjugated | |
SLCO2A1 | |
OATP2A1, PGT, Prostaglandin transporter, SLC21A2, SLCO2A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24059, 6578 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92959, Q9EPT5 | |
SLCO2A1 | |
PGT Fusion Protein Ag5594 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |