Läs mer
SI Analytics™ pH-kombinationselektrod med 1m fast kabel
Handla allt SI Analytics Produkter
Kontakttyper:
BNC
DIN, 2 mm stiftkontakt, 2 mm Pin Plug
DÅN
Fast kabel, 4 mm Banana Plug
Korsning:
Keramiskt membran
Platinamembran
Förpackningsstorlek:
Varje
Beskrivning
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Specifikationer
Specifikationer
| Kontakttyper | BNC |
| Korsning | Platinamembran |
| Material | A-Glass |
| pH-intervall | 0 to 14 |
| Typ | Combination pH Electrode |
| Intern referens | Silamid™ |
| Membran | Sphere |
| Modell | N 52 BNC |
| Temperaturområde (metriskt). | -5°C to +100°C |
| Längd (metrisk) | 120 mm |
| Visa mer |