Learn More
Fisherbrand™ pH/Temp Electrode with BNC Connector
Ideal for loaded or viscous solutions, the pHast Clean electrode increases the life of the electrode and reduces cleaning operations.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- The pHast Clean electrode makes it possible to renew the junction by forcing the passage of the electrolyte through pressure on the electrode head
- This system increases the life of the electrode, reduces cleaning operations and measured
- Ideal for loaded or viscous solutions
- Electrolyte KCl 4M + AgCl
Specifications
BNC | |
pH Electrode | |
13 |
12 mm | |
120 mm | |
-5°C to +100°C |