Learn More
Fisherbrand™ pH/Temp Electrode with BNC Connector
Ideal for loaded or viscous solutions, the pHast Clean electrode increases the life of the electrode and reduces cleaning operations.
Brand:  Fisherbrand™
Additional Details : Weight : 0.80000kg
Product Code. 11729798
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • The pHast Clean electrode makes it possible to renew the junction by forcing the passage of the electrolyte through pressure on the electrode head
  • This system increases the life of the electrode, reduces cleaning operations and measured
  • Ideal for loaded or viscous solutions
  • Electrolyte KCl 4M + AgCl
Specifications
Specifications
BNC
pH Electrode
13
12 mm
120 mm
-5°C to +100°C