Learn More
WTW™ pH Test Kits
Brand: WTW™ 252050
Additional Details : Weight : 0.80000kg
Specifications
ISO 6439 | |
2 mL | |
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer | |
15°C to 25°C | |
Nitrate (NO3) Cuvette Test Kit |
<1 at 20°C | |
Ground water, drinking water, surface water, swimming pools | |
Nitrate | |
Chromotrope acid |