Läs mer
Tocris Bioscience™ PHA 543613 hydrochloride
Beskrivning
Potent α7 nAChR agonist that displays selectivity over α3β4, α1β1γδ, α4β2 and 5-HT3 receptors. Positively influences sensory gating and memory in in vivo models of schizophrenia. Orally active and brain penetrant.
Specifikationer
Specifikationer
| Kemiskt namn eller material | PHA 543613 hydrochloride |
| CAS | 478149-53-0 |
| Kvantitet | 10 mg |
| Molekylformel | C15H18ClN3O2 |
| Formel vikt | 307.78 |
| Renhet | >98% |
| Molekylvikt (g/mol) | 307.778 |