Läs mer
PHA-767491 hydrochloride, MedChemExpress
Produktkod. 16481857
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16481857 50 mg
16465698 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16481857 Leverantör Medchem Express Leverantörsnummer HY13461A
MedChemExpress PHA-767491 hydrochloride is a dual Cdc7/Cdk9 inhibitor, with IC50s of 10 nM and 34 nM, respectively.
PHA-767491 hydrochloride is a dual Cdc7/Cdk9 inhibitor, with IC50s of 10 nM and 34 nM, respectively.
Specifikationer
Kemiskt namn eller material PHA-767491 hydrochloride
CAS 942425-68-5
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C12H12ClN3O
Kvantitet 50 mg
Synonym CAY-10572 hydrochloride
Löslighetsinformation H2O : 50 mg/mL (200.24 mM; Need ultrasonic) ∣DMSO : 17.33 mg/mL (69.40 mM; Need ultrasonic and warming)
LEDER O=C1C2=C(NC(C3=CC=NC=C3)=C2)CCN1.[H]Cl
Molekylvikt (g/mol) 249.7
Formel vikt 249.7
Procent renhet 99.91%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease