Läs mer
Phalloidine, TRC
Produktkod. 30428600
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30428600 1 mg
1 options
Produktkod. 30428600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2945231MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phalloidine
CAS 17466-45-4
Molekylformel C35 H48 N8 O11 S
Kvantitet 1 mg
Synonym Cyclo[L-alanyl-D-threonyl-L-cysteinyl-(4S)-4-hydroxy-L-prolyl-L-alanyl-2-mercapto-L-tryptophyl-(4R)-4,5-dihydroxy-L-leucyl] Cyclic (3→6)-thioether, 18,9-(Iminoethaniminoethaniminoethaniminomethano)-17H-pyrrolo[1',2':5,6][1,5,8,11]thiatriazacyclopentadecino[15,14-b]indole Cyclic Peptide Derivative, NSC 523214
InChI formel InChI=1S/C35H48N8O11S/c1-15-27(47)38-22-10-20-19-7-5-6-8-21(19)41-33(20)55-13-24(34(53)43-12-18(46)9-25(43)31(51)37-15)40-32(52)26(17(3)45)42-28(48)16(2)36-30(50)23(39-29(22)49)11-35(4,54)14-44/h5-8,15-18,22-26,41,44-46,54H,9-14H2,1-4H3,(H,36,50)(H,37,51)(H,38,47)(H,39,49)(H,40,52)(H,42,48)/t15-,16-,17-,18-,22-,23-,24?,25-,26+,35+/m0/s1
LEDER C[C@H](O)[C@H]1NC(=O)[C@H](C)NC(=O)[C@H](C[C@@](C)(O)CO)NC(=O)[C@@H]2Cc3c(SC[C@H](NC1=O)C(=O)N4C[C@@H](O)C[C@H]4C(=O)N[C@@H](C)C(=O)N2)[nH]c5ccccc35
Molekylvikt (g/mol) 788.87
Formel vikt 788.3163