Läs mer
Kern Sohn Phase Contrast Microscope OBL
Beskrivning
- We have developed this series specially for general applications with phase contrast method. In addition, the stable, modular construction system of the OBL series offers many more options.
- A strong and continously adjustable 20 W halogen illumination unit (Philips) ensures the optimum lighting conditions
- A special fixed, pre-centred phase contrast condenser with aperture diaphragm as well as field diaphragm give you a simplified Koehler illumination and thereby a powerful phase-contrast display of your sample.
- The large mechanical stage and its specimen holder holds up to two samples at the same time and is quick and easy to focus using a coaxial coarse and fine focusing knob on both sides
- A large selection of eyepieces, objectives and colour filters, a simple polarizing unit as well as further phase contrast units are available to you as accessories
- A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
- Scope of application: haematology, urology, gynaecology, dermatology, pathology, microbiology and parasitology, immunology, Sewage treatment plants, oncology, entomology, Veterinary, water analysis, breweries
- Applications/Samples: Specially for extremely translucent, thin, low-contrast, challenging samples (e.g. living mammal cells, bacteria, tissue) with phase contrast
Specifikationer
Specifikationer
| Huvud stil | Trinokulär |
| Typ av belysning | Halogen |
| Förstoringskraft | 40X, 10X, 40X, 100X |
| Spänning | 230 V |