Learn More
PHC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561504
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PQLASVSPSV ALQPSSEAHA MPLGPVTPAL PLQCPTANLH KPGGSQQCHP PTPDTGPQNG HPEGVPHTPQ RRFQHTSAVI LQLQPASPVP QQCVPDDWKE VAPGEKSVPE TRSGPSPHQQ AIVTAMP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 83%.
In Drosophila melanogaster, the 'Polycomb' group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein 'polyhomeotic' (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.Specifications
PHC2 | |
Polyclonal | |
Unconjugated | |
Phc2 | |
A3galt2; AA415044; D130050K19Rik; D4Ertd810e; early development regulator 2; early development regulator 2 (homolog of polyhomeotic 2); early development regulatory protein 2; Edr2; hPH2; mPH2; p36; Ph2; Phc2; polyhomeotic 2; polyhomeotic 2 protein; polyhomeotic homolog 2; polyhomeotic homolog 2 (Drosophila); polyhomeotic-like 2; polyhomeotic-like 2 (Drosophila); polyhomeotic-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXK0 | |
Phc2 | |
Recombinant protein corresponding to Human PHC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only