Läs mer
Phenaglycodol, TRC
Produktkod. 30411333
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411333 50 mg
30424330 10 mg
30426174 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29472550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 79-93-6
Molekylformel C11H15ClO2
Molekylvikt (g/mol) 214.69
Synonym 2-(4-Chlorophenyl)-3-methyl-2,3-butanediol, 2-p-Chlorophenyl-3-methyl-2,3-butanediol, Acalmid, Acalo, Alterton, Felixyn, NSC 169451, NSC 63894, Sinforil, Stesil, Ultran
IUPAC-namn 2-(4-chlorophenyl)-3-methylbutane-2,3-diol
LEDER CC(C)(O)C(C)(O)c1ccc(Cl)cc1
Specifikationer
Kemiskt namn eller material Phenaglycodol
Kvantitet 50 mg
InChI formel InChI=1S/C11H15ClO2/c1-10(2,13)11(3,14)8-4-6-9(12)7-5-8/h4-7,13-14H,1-3H3
Formel vikt 214.0761