Learn More
Phenanthrene-9,10-13C2, TRC
Product Code. 30425550
Change view
Click to view available options
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30425550 1 mg
30423140 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30425550 Supplier Toronto Research Chemicals Supplier No. TRCP2948041MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 334973-64-7
Molekylformel C1213C2H10
Molekylvikt (g/mol) 180.21
Synonym [3]Helicene-9,10-13C2, NSC 26256-9,10-13C2, Ravatite-9,10-13C2
IUPAC-namn phenanthrene
LEDER C12=CC=CC=C1C3=C(C=CC=C3)[13CH]=[13CH]2
Specifications
Kemiskt namn eller material Phenanthrene-9,10-13C2
Kvantitet 1 mg
InChI formel InChI=1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H/i9+1,10+1
Formel vikt 180.085