missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenazine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 174.0°C to 177.0°C |
Color | Yellow to Brown |
Boiling Point | >360.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Beilstein | 23,223 |
Merck Index | 15,7331 |
Solubility Information | Solubility in water: insoluble. Other solubilities: partially soluble in alcohol and ether, moderately soluble in benzene |
Show More |
Safety and Handling
Product Identifier
- Phenazine
- Warning
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'