Läs mer
Phenazopyridine Hydrochloride, TRC
Produktkod. 30136917
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136917 100 mg
30132669 1 g
30011012 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP313750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136-40-3
Molekylformel C11 H11 N5 . Cl H
Molekylvikt (g/mol) 249.7
Synonym 2,6-Pyridinediamine, 3-(2-phenyldiazenyl)-, hydrochloride (1:1), 2,6-Pyridinediamine, 3-(phenylazo)-, monohydrochloride (9CI), Pyridine, 2,6-diamino-3-(phenylazo)-, monohydrochloride (8CI), 2,6-Diamino-3-phenylazopyridine hydrochloride, 2,6-Diamino-3-phenylazopyridine monohydrochloride, Azodyne, Bisteril, Diridone, Mallophene, NC 150, Phenazodine, Phenazopyridine hydrochloride, Phenazopyridinium chloride, Phenylazo tablets, Pirid, Pyridacil, Pyridiate, Pyridium, Pyripyridium, Sedural, Uridinal, Urisept, Urodine, W 1655, β-Phenylazo-α,α'-diaminopyridine hydrochloride
IUPAC-namn 3-[(E)-phenyldiazenyl]pyridine-2,6-diamine;hydrochloride
LEDER Cl.Nc1ccc(N=Nc2ccccc2)c(N)n1
Specifikationer
Kemiskt namn eller material Phenazopyridine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C11H11N5.ClH/c12-10-7-6-9(11(13)14-10)16-15-8-4-2-1-3-5-8;/h1-7H,(H4,12,13,14);1H
Formel vikt 249.0781