Läs mer
Phenazopyridine, TRC
Produktkod. 30413554
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413554 100 mg
30417965 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP313751100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 94-78-0
Molekylformel C11H11N5
Molekylvikt (g/mol) 213.24
Synonym 3-(2-Phenyldiazenyl)-2,6-pyridinediamine Hydrochloride, 3-(Phenylazo)-2,6-pyridinediamine, 2,6-Diamino-3-phenylazopyridine, Gastracid, Gastrotest, NSC 145895
IUPAC-namn 3-[(E)-phenyldiazenyl]pyridine-2,6-diamine
LEDER Nc1ccc(N=Nc2ccccc2)c(N)n1
Specifikationer
Kemiskt namn eller material Phenazopyridine
Kvantitet 100 mg
InChI formel InChI=1S/C11H11N5/c12-10-7-6-9(11(13)14-10)16-15-8-4-2-1-3-5-8/h1-7H,(H4,12,13,14)
Formel vikt 213.1014