Läs mer
Phenelzine Sulfate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phenelzine Sulfate |
| CAS | 156-51-4 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C8 H12 N2 . H2 O4 S |
| Kvantitet | 500 mg |
| Synonym | Hydrazine','(2-phenylethyl)-','sulfate (1:1), Hydrazine','phenethyl-','sulfate (1:1) (8CI), (2-Phenylethyl)hydrazine dihydrogen sulfate, (β-Phenylethyl)hydrazine dihydrogen sulfate, (β-Phenylethyl)hydrazine sulfate, 2-Phenethylhydrazine sulfate, Estinerval, Fenelzin, Kalgan, Mao-rem, NSC 170957, Nardelzine, Nardil, Phenalzine dihydrogen sulfate, Phenelzine acid sulfate, Phenelzine bisulfate, Phenelzine dihydrogen sulfate, Phenelzine hydrogen sulfate, Phenelzine sulfate, Phenethylhydrazine sulfate, Stinerval, W 1544A |
| InChI formel | InChI=1S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10H,6-7,9H2;(H2,1,2,3,4) |
| LEDER | NNCCc1ccccc1.OS(=O)(=O)O |
| IUPAC-namn | 2-phenylethylhydrazine;sulfuric acid |
| Molekylvikt (g/mol) | 234.27 |
| Visa mer |