missing translation for 'onlineSavingsMsg'
Läs mer

Phenethicillin Sodium Salt, TRC

Produktkod. 30420234
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420234 10 mg
30411309 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30420234 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29632010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 30302-52-4
Molekylformel C17H19N2NaO5S
Molekylvikt (g/mol) 386.4
Synonym Pheneticillin Sodium Salt, Phenoxy PC Sodium Salt, Phenoxyethylpenicillin Sodium Salt, Synthepen Sodium Salt, α-Phenoxyethylpenicillin Sodium Salt, 1-Phenoxyethylpenicillin Sodium Salt, Alfacilin Sodium Salt, Feneticillin Sodium Salt, NSC 117548 Sodium Salt, Optipen Sodium Salt, Penicillin 152 Sodium Salt, Penicillin MV Sodium Salt, Peniplus Sodium Salt, Penorale Sodium Salt, Penova Sodium Salt, (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[(1-oxo-2-phenoxypropyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid Sodium Salt
LEDER O=C1[C@@H](NC(C(OC2=CC=CC=C2)C)=O)C3N1[C@@H](C(O[Na])=O)C(C)(C)S3

Specifikationer

Kemiskt namn eller material Phenethicillin Sodium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C17H20N2O5S.Na/c1-9(24-10-7-5-4-6-8-10)13(20)18-11-14(21)19-12(16(22)23)17(2,3)25-15(11)19;/h4-9,11-12,15H,1-3H3,(H,18,20)(H,22,23);/q;+1/p-1/t9?,11-,12+,15?;/m1./s1
Formel vikt 386.0912
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.