Learn More
Phenetole, 98+%, Thermo Scientific Chemicals
Catalog Number p-180915
Click to view available options
Quantity:
100 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
103-73-1
C8H10O
122.17
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1
103-73-1
C8H10O
122.17
MFCD00009090
DLRJIFUOBPOJNS-UHFFFAOYSA-N
phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
7674
CHEBI:67129
ethoxybenzene
CCOC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC2H5
Refractive Index 1.5055 to 1.5075
Quantity 100 mL
Beilstein 06, 140
Merck Index 15, 7343
Specific Gravity 0.96
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and carbon tetrachloride
Formula Weight 122.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Phenetole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Phenetole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement