Läs mer
Phenformin Hydrochloride, TRC
Produktkod. 30423427
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423427 10 g
30423794 5 g
30410304 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30423427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29690010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 834-28-6
Molekylformel C10 H15 N5 . Cl H
Molekylvikt (g/mol) 241.72
Synonym Imidodicarbonimidic diamide, N-(2-phenylethyl)-, hydrochloride (1:1), Biguanide, 1-phenethyl-, monohydrochloride (8CI), Imidodicarbonimidic diamide, N-(2-phenylethyl)-, monohydrochloride (9CI), 1-(2-Phenylethyl)biguanide hydrochloride, 1-Phenethylbiguanide hydrochloride, 1-β-Phenylethylbiguanide hydrochloride, Azucaps, DBI-TD, Debeone DT, Debinyl, Dibein, Dipar, Feguanide, Glucopostin, Insoral, Lentobetic, Meltrol, N-Phenethylbiguanide hydrochloride, Normoglucina, N'-β-Phenylethylbiguanide-hydrochloride, Phenethylbiguanide hydrochloride, Phenformin hydrochloride, α-Phenethylbiguanide hydrochloride
IUPAC-namn 1-carbamimidoyl-3-(2-phenylethyl)guanidine;hydrochloride
LEDER Cl.NC(=N)NC(=N)NCCc1ccccc1
Specifikationer
Kemiskt namn eller material Phenformin Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C10H15N5.ClH/c11-9(12)15-10(13)14-7-6-8-4-2-1-3-5-8;/h1-5H,6-7H2,(H6,11,12,13,14,15);1H
Formel vikt 241.1094
Procent renhet >95%