Läs mer
Pheniramine 1.0 mg/ml in Methanol, LoGiCal
Produktkod. 30459164
Change view
Kvantitet:
1 ml
Produktkod. Kvantitet
30459164 1 ml
1 options
Produktkod. 30459164 Leverantör LoGiCal Leverantörsnummer LGCAMP0059.0101
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
Pheniramine 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pheniramine
CAS 86-21-5
Analyt- eller komponentnamn Pheniramine
Frakt skick Room Temperature
Molekylformel C16 H20 N2
Certifikat och efterlevnad ISO 17034
Synonym Brompheniramine Maleate Imp. C (EP), Brompheniramine Imp. C (EP), (3 RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine, Pheniramine, Brompheniramine Maleate Impurity C, Brompheniramine Impurity C
InChI formel InChI=1 S/C16H20N2/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16/h3-10,12,15 H,11,13H2,1-2H3
LEDER CN(C)CCC(c1ccccc1)c2ccccn2
IUPAC-namn N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine
Lösningsmedel eller matris Methanol
Industry Type Forensics
Molekylvikt (g/mol) 240.34
Kvantitet 1 ml
Formel vikt 240.1626 g/mol