Läs mer
Pheniramine Maleate, TRC
Produktkod. 30133519
Change view
Kvantitet:
50 mg
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133519 50 g
17931455 50 mg
17943565 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133519 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29720050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 132-20-7
Molekylformel C16 H20 N2 . C4 H4 O4
Molekylvikt (g/mol) 356.42
Synonym (3RS)-N,N-Dimethyl-3-phenyl-3-(pyridin-2-yl)propan-1-amine Maleate, Pheniramine Maleate, Dexchlorpheniramine Maleate Imp. A (EP) as Maleate, Brompheniramine Maleate Imp. C (EP) as Maleate, N,N-Dimethyl-γ-phenyl-2-pyridinepropanamine (2Z)-2-butenedioate (1:1), N,N-Dimethyl-γ-phenyl-2-pyridinepropanamine (Z)-2-butenedioate (1:1), Prophenpyridamine maleate, 2-[α-[2-(Dimethylamino)ethyl]benzyl]pyridine maleate (1:1), 2-Pyridinepropanamine, N,N-dimethyl-γ-phenyl-, (2Z)-2-butenedioate (1:1) (9CI, ACI), 2-Pyridinepropanamine, N,N-dimethyl-γ-phenyl-, (Z)-2-butenedioate (1:1) (ZCI), Prophenpyridamine maleate (6CI), Pyridine, 2-[α-[2-(dimethylamino)ethyl]benzyl]-, maleate (1:1) (8CI), 1-Phenyl-1-(2-pyridyl)-3-dimethaminopropane maleinate, 1-Phenyl-1-(2-pyridyl)-3-dimethylaminopropane maleate, 2-[α-[2-(Dimethylamino)ethyl]benzyl]pyridine bimaleate, Antolozine, Daneral, Inhiston, Metron, Naphcon A, Pheniramine hydrogen maleate, Pheniramine maleate, PM 241, Trimeton, Trimetose
IUPAC-namn (Z)-but-2-enedioic acid;N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine
LEDER CN(C)CCC(c1ccccc1)c2ccccn2.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Pheniramine Maleate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 g
InChI formel InChI=1S/C16H20N2.C4H4O4/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16;5-3(6)1-2-4(7)8/h3-10,12,15H,11,13H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vikt 356.1736
Procent renhet >95%