missing translation for 'onlineSavingsMsg'
Läs mer

Phenol-d5, TRC

Produktkod. 30417044
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417044 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30417044 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3180031G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Phenol-d5
CAS 4165-62-2
Anteckningar om renhetsgrad GC
Molekylformel C6 D5 H O
Kvantitet 1 g
Synonym Phenol-2,3,4,5,6 D5, Phenol (ring-D5), Phen-2,3,4,5,6-d5-ol, Phen-d5-ol (7CI,8CI,9CI), 2,3,4,5,6-Pentadeuteriophenol, Phenol D5
InChI formel InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,2D,3D,4D,5D
LEDER [2H]c1c([2H])c([2H])c(O)c([2H])c1[2H]
IUPAC-namn 2,3,4,5,6-Pentadeuteriophenol
Molekylvikt (g/mol) 99.14
Formel vikt 99.0732
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.