missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol USP Crystal, Macron Fine Chemicals™
Click to view available options
Quantity:
2.5 kg
12 x 500 g
Packaging:
Glass
Poly Coated
Specifications
Packaging | Poly Coated |
Quantity | 2.5 kg |
Formula Weight | 94.11 |
Grade | USP |
Physical Form | Crystalline |
Chemical Name or Material | Phenol |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'