Läs mer
Phenolphthalein b-D-Glucuronide (~90%), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phenolphthalein beta-D-Glucuronide |
| CAS | 15265-26-6 |
| Molekylformel | C26H22O10 |
| Kvantitet | 25 mg |
| Synonym | 4-[1,3-Dihydro-1-(4-hydroxyphenyl)-3-oxo-1-isobenzofuranyl]phenyl beta-D-Glucopyranosiduronic Acid, p-[3-(p-Hydroxyphenyl)phthalidyl]phenyl beta-D-Glucosiduronic Acid, Phenolphthalein Glucuronide, Phenolphthalein mono-beta-Glucuronide, Phenolphthalein Monoglucuronide, Phenolphthalein-4'-beta-D-glucopyranosiduronide, Phenolphthalein-beta-D-glucosiduronate, Phenolphthalein-beta-glucuronide |
| InChI formel | InChI=1S/C26H22O10/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32/h1-12,19-22,25,27-30H,(H,31,32)/t19-,20-,21+,22-,25+,26?/m0/s1 |
| LEDER | O[C@H]1[C@H](Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O |
| IUPAC-namn | (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[1-(4-hydroxyphenyl)-3-oxo-2-benzofuran-1-yl]phenoxy]oxane-2-carboxylic acid |
| Molekylvikt (g/mol) | 494.45 |
| Formel vikt | 494.1213 |