missing translation for 'onlineSavingsMsg'
Läs mer

Phenolphthalein b-D-Glucuronide (~90%), TRC

Produktkod. 30417420
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417420 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30417420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31801550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Phenolphthalein beta-D-Glucuronide
CAS 15265-26-6
Molekylformel C26H22O10
Kvantitet 50 mg
Synonym 4-[1,3-Dihydro-1-(4-hydroxyphenyl)-3-oxo-1-isobenzofuranyl]phenyl beta-D-Glucopyranosiduronic Acid, p-[3-(p-Hydroxyphenyl)phthalidyl]phenyl beta-D-Glucosiduronic Acid, Phenolphthalein Glucuronide, Phenolphthalein mono-beta-Glucuronide, Phenolphthalein Monoglucuronide, Phenolphthalein-4'-beta-D-glucopyranosiduronide, Phenolphthalein-beta-D-glucosiduronate, Phenolphthalein-beta-glucuronide
InChI formel InChI=1S/C26H22O10/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32/h1-12,19-22,25,27-30H,(H,31,32)/t19-,20-,21+,22-,25+,26?/m0/s1
LEDER O[C@H]1[C@H](Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[1-(4-hydroxyphenyl)-3-oxo-2-benzofuran-1-yl]phenoxy]oxane-2-carboxylic acid
Molekylvikt (g/mol) 494.45
Formel vikt 494.1213
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.