Läs mer
Phenolphthalein Disodium Salt (>90%), TRC
Produktkod. 30418225
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418225 500 mg
30428584 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30418225 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP318033500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenolphthalein Disodium Salt
CAS 518-51-4
Molekylformel C20H12Na2O4
Kvantitet 500 mg
InChI formel InChI=1S/C20H14O4.2Na/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20;;/h1-12,21-22H;;/q;2*+1/p-2
LEDER [Na+].[Na+].[O-]c1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc([O-])cc4
IUPAC-namn disodium;4-[1-(4-oxidophenyl)-3-oxo-2-benzofuran-1-yl]phenolate
Molekylvikt (g/mol) 362.29
Formel vikt 362.0531