missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, U.S.P./N.F. grade, MP Biomedicals™

Catalog Number p-4679915
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
This item is not returnable. View return policy
77-09-8
C20H14O4
318.33
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone
4764
CHEBI:34914
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Assay Percent Range U.S.P./N.F. grade
Formula Weight 318.3
Density 1.29g/cm3
Color White to White/Yellow
Quantity 100 g
Physical Form Crystals
Chemical Name or Material Phenolphthalein
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'