Läs mer
Phenoxymethylpenilloic Acid (Mixture of Diastereomers), TRC
Produktkod. 30011023
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011023 100 mg
30006568 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011023 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP298250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4847-29-4
Molekylformel C15 H20 N2 O4 S
Molekylvikt (g/mol) 324.4
Synonym 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-, 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[(2-phenoxyacetamido)methyl]- (6CI,7CI,8CI), 4-Thiazolidinecarboxylic acid, 5,5-dimethyl-2-[[(phenoxyacetyl)amino]methyl]- (9CI), 5,5-Dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-4-thiazolidinecarboxylic acid, Penilloic acid V, Penilloic acid, phenoxymethyl-, Phenoxymethylpenilloic acid, 5,5-Dimethyl-2-[[(phenoxyacetyl)amino]methyl]thiazolidine-4-carboxylic acid
IUPAC-namn 5,5-dimethyl-2-[[(2-phenoxyacetyl)amino]methyl]-1,3-thiazolidine-4-carboxylic acid
LEDER CC1(C)SC(CNC(=O)COc2ccccc2)NC1C(=O)O
Specifikationer
Kemiskt namn eller material Phenoxymethylpenilloic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C15H20N2O4S/c1-15(2)13(14(19)20)17-12(22-15)8-16-11(18)9-21-10-6-4-3-5-7-10/h3-7,12-13,17H,8-9H2,1-2H3,(H,16,18)(H,19,20)
Formel vikt 324.1144