Läs mer
Phenoxypropazine Dihydrochloride, LoGiCal
Produktkod. 30462009
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462009 10 mg
1 options
Produktkod. 30462009 Leverantör LoGiCal Leverantörsnummer LGCFOR3185.02
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenoxypropazine Dihydrochloride
CAS 1803608-13-0
Analyt- eller komponentnamn Phenoxypropazine Dihydrochloride
Frakt skick Room Temperature
Molekylformel C9 H14 N2 O . 2 Cl H
Certifikat och efterlevnad ISO 9001
Synonym Prochlorperazine N1,N4,S,S-Tetraoxide Dihydrochloride
InChI formel InChI=1S/C9H14N2O.2ClH/c1-8(11-10)7-12-9-5-3-2-4-6-9;;/h2-6,8,11H,7,10H2,1H3;2*1H
Lösningstyp Neat
LEDER Cl.Cl.CC(COc1ccccc1)NN
IUPAC-namn 1-phenoxypropan-2-ylhydrazine;dihydrochloride
Industry Type Food Testing
Molekylvikt (g/mol) 239.14
Kvantitet 10 mg
Formel vikt 238.064 g/mol