Läs mer
Phenprocoumon, TRC
Produktkod. 30420169
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420169 5 mg
30415502 10 mg
30409314 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420169 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3288205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 435-97-2
Molekylformel C18 H16 O3
Molekylvikt (g/mol) 280.32
Synonym 3-(alpha-Ethylbenzyl)-4-hydroxycoumarin, (+/-)-Phenprocoumon, 3-(1-Phenylpropyl)-4-hydroxycoumarin, 3-(alpha-Phenylpropyl)-4-hydroxycoumarin, 4-Hydroxy-2-oxo-3-(1-phenylpropyl)-2H-chromene, DL-3-(alpha-Ethylbenzyl)-4-hydroxycoumarin, Falithrom, Fencumar, Liquamar, Marcoumar, Marcumar, Phenprocoumarol, Phenprocoumarole, Phenprocoumon
IUPAC-namn 4-hydroxy-3-(1-phenylpropyl)chromen-2-one
LEDER CCC(C1=C(O)c2ccccc2OC1=O)c3ccccc3
Specifikationer
Kemiskt namn eller material Phenprocoumon
Kvantitet 5 mg
InChI formel InChI=1S/C18H16O3/c1-2-13(12-8-4-3-5-9-12)16-17(19)14-10-6-7-11-15(14)21-18(16)20/h3-11,13,19H,2H2,1H3
Formel vikt 280.1099