Läs mer
Phenyl(2-pyridyl)methanol, Mikromol™
Produktkod. 30049172
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049172 25 mg
17963326 100 mg
30459931 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049172 Leverantör Mikromol Leverantörsnummer MM0101.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phenyl(2-pyridyl)methanol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl(2-pyridyl)methanol
CAS 14159-57-0
Analyt- eller komponentnamn Phenyl(2-pyridyl)methanol
Frakt skick Room Temperature
Molekylformel C12 H11 N O
Certifikat och efterlevnad ISO 17025
Synonym NSC 241060, Phenyl(2-pyridinyl)methanol, Phenyl(2-pyridyl)methanol, Phenyl-2-pyridylcarbinol, alpha-Phenyl-2-pyridinemethanol, (+/-)-2-(alpha-Hydroxybenzyl)pyridine, (+/-)-2-Pyridylphenylmethanol, 2-(alpha-Hydroxybenzyl)pyridine
InChI formel InChI=1S/C12H11NO/c14-12(10-6-2-1-3-7-10)11-8-4-5-9-13-11/h1-9,12,14H
LEDER OC(c1ccccc1)c2ccccn2
IUPAC-namn phenyl(pyridin-2-yl)methanol
Molekylvikt (g/mol) 185.22
Kvantitet 25 mg
Formel vikt 185.0841 g/mol