Läs mer
Phenyl 3,4-Di-O-acetyl-Alpha-O-benzyl-1-thio-Alpha-L-rhamnopyranoside, TRC
Produktkod. 30414858
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414858 25 mg
30409825 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414858 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31970525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl 3,4-Di-O-acetyl-a-O-benzyl-1-thio-a-L-rhamnopyranosid
CAS 849938-20-1
Molekylformel C23 H26 O6 S
Kvantitet 25 mg
Synonym Phenyl 3,4-Di-O-Acetyl-α-O-Benzyl-1-Thio-α-L-Rhamnopyranoside
InChI formel InChI=1S/C23H26O6S/c1-15-20(28-16(2)24)21(29-17(3)25)22(26-14-18-10-6-4-7-11-18)23(27-15)30-19-12-8-5-9-13-19/h4-13,15,20-23H,14H2,1-3H3/t15-,20-,21+,22+,23-/m0/s1
LEDER C[C@@H]1O[C@@H](Sc2ccccc2)[C@H](OCc3ccccc3)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC-namn [(2S,3S,4R,5R,6S)-4-acetyloxy-2-methyl-5-phenylmethoxy-6-phenylsulfanyloxan-3-yl] acetate
Molekylvikt (g/mol) 430.51
Formel vikt 430.145