Läs mer
Phenyl 3-Desethyl 3-Propyl Carbodenafil, TRC
Produktkod. 30135888
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135888 100 mg
30005554 10 mg
30012622 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP416565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2711006-61-8
Molekylformel C29 H34 N6 O3
Molekylvikt (g/mol) 514.63
Synonym Carbodenafil impurity 3, 1-Methyl-5-(5-(4-phenylpiperazine-1-carbonyl)-2-propoxyphenyl)-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
IUPAC-namn 1-methyl-5-[5-(4-phenylpiperazine-1-carbonyl)-2-propoxy-phenyl]-3-propyl-4~{H}-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCOc1ccc(cc1C2=NC(=O)c3c(N2)c(CCC)nn3C)C(=O)N4CCN(CC4)c5ccccc5
Specifikationer
Kemiskt namn eller material Phenyl 3-Desethyl 3-Propyl Carbodenafil
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C29H34N6O3/c1-4-9-23-25-26(33(3)32-23)28(36)31-27(30-25)22-19-20(12-13-24(22)38-18-5-2)29(37)35-16-14-34(15-17-35)21-10-7-6-8-11-21/h6-8,10-13,19H,4-5,9,14-18H2,1-3H3,(H,30,31,36)
Formel vikt 514.2692
Procent renhet >95%