Läs mer
Phenyl 4-Methoxybenzoate, TRC
Produktkod. 30409866
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30409866 50 mg
1 options
Produktkod. 30409866 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP57166050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl 4-Methoxybenzoate
CAS 4181-97-9
Molekylformel C14 H12 O3
Kvantitet 50 mg
Synonym 4-Methoxybenzoic Acid Phenyl Ester
InChI formel InChI=1S/C14H12O3/c1-16-12-9-7-11(8-10-12)14(15)17-13-5-3-2-4-6-13/h2-10H,1H3
LEDER COc1ccc(cc1)C(=O)Oc2ccccc2
IUPAC-namn phenyl 4-methoxybenzoate
Molekylvikt (g/mol) 228.24
Formel vikt 228.0786