Läs mer
Phenyl b-D-Glucuronide, TRC
Produktkod. 30418355
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30418355 1 g
1 options
Produktkod. 30418355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3265001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl beta-D-Glucuronide
CAS 17685-05-1
Molekylformel C12 H14 O7
Kvantitet 1 g
Synonym Phenol-Glucuronide
InChI formel InChI=1S/C12H14O7/c13-7-8(14)10(11(16)17)19-12(9(7)15)18-6-4-2-1-3-5-6/h1-5,7-10,12-15H,(H,16,17)/t7-,8-,9+,10-,12+/m0/s1
LEDER O[C@@H]1[C@@H](O)[C@H](Oc2ccccc2)O[C@@H]([C@H]1O)C(=O)O
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 270.24
Formel vikt 270.074