Läs mer
Phenyl Beta-D-Glucuronide Methyl Ester, TRC
Produktkod. 30425715
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30425715 250 mg
1 options
Produktkod. 30425715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP326525250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl beta-D-Glucuronide Methyl Ester
CAS 55811-42-2
Molekylformel C13H16O7
Kvantitet 250 mg
Synonym (2S,3S,4S,5R,6S)-Methyl 3,4,5-Trihydroxy-6-phenoxytetrahydro-2H-pyran-2-carboxylate
InChI formel InChI=1S/C13H16O7/c1-18-12(17)11-9(15)8(14)10(16)13(20-11)19-7-5-3-2-4-6-7/h2-6,8-11,13-16H,1H3/t8-,9-,10+,11-,13+/m0/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2ccccc2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxane-2-carboxylate
Molekylvikt (g/mol) 284.26
Formel vikt 284.0896