Läs mer
Phenyl beta-D-Thioglucopyranoside, TRC
Produktkod. 30415231
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30415231 5 g
1 options
Produktkod. 30415231 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3265505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenyl beta-D-Thioglucopyranoside
CAS 2936-70-1
Molekylformel C12H16O5S
Kvantitet 5 g
Synonym β-D-Phenyl 1-Thio-Glucopyranoside, Phenyl 1-Thio-Glucopyranoside, NSC 231833
InChI formel InChI=1S/C12H16O5S/c13-6-8-9(14)10(15)11(16)12(17-8)18-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9-,10+,11-,12+/m1/s1
LEDER O[C@H]([C@H]([C@@H]([C@H](O1)CO)O)O)[C@@H]1SC2=CC=CC=C2
IUPAC-namn (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol
Molekylvikt (g/mol) 272.32
Formel vikt 272.07