missing translation for 'onlineSavingsMsg'
Learn More

Phenyl bromoacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7040644
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr
This item is not returnable. View return policy
620-72-4
C8H7BrO2
215.046
MFCD00192391
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
564919
phenyl 2-bromoacetate
C1=CC=C(C=C1)OC(=O)CBr

Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

Solubility
Soluble in ethanol and ether, insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 31°C to 33°C
Density 1.508
Boiling Point 134°C
Flash Point 113°C (235°F)
Linear Formula BrCH2CO2C6H5
Quantity 5 g
Solubility Information Soluble in ethanol and ether,insoluble in water.
Formula Weight 215.04
Percent Purity 98%
Chemical Name or Material Phenyl bromoacetate
compliance-icons
Product Identifier
  • Phenyl bromoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Bromo-acetic acid phenyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'