missing translation for 'onlineSavingsMsg'
Learn More

Phenyl chlorothionocarbonate, 99%, Thermo Scientific Chemicals

Catalog Number p-4509973
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1005-56-7
C7H5ClOS
172.63
MFCD00004920
KOSYAAIZOGNATQ-UHFFFAOYSA-N
o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate
70498
O-phenyl chloromethanethioate
ClC(=S)OC1=CC=CC=C1
This item is not returnable. View return policy
1005-56-7
C7H5ClOS
172.63
MFCD00004920
KOSYAAIZOGNATQ-UHFFFAOYSA-N
o-phenyl carbonochloridothioate, phenyl chlorothionoformate, o-phenyl chlorothioformate, phenyl chlorothioformate, phenyl thioxochloroformate, phenyl chlorothionocarbonate, phenoxythiocarbonyl chloride, chlorothioformic acid phenyl ester, o-phenyl chlorothionoformate, o-phenyl chlorothiocarbonate
70498
O-phenyl chloromethanethioate
ClC(=S)OC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.2480g/mL
Boiling Point 81.0°C to 83.0°C (6.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC(S)OC6H5
Refractive Index 1.4795 to 1.4815
Quantity 1 g
Beilstein 06,III,609
Fieser 10,306
Specific Gravity 1.248
Solubility Information Solubility in water: decomposes
Formula Weight 172.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenyl chlorothionocarbonate
Show More Show Less
compliance-icons
Product Identifier
  • Phenyl chlorothionocarbonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'