Läs mer
Phenyl Cyanate (Stabilized with PPE), TRC
Produktkod. 30425401
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425401 5 g
30425888 1 g
30425662 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425401 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3196755G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1122-85-6
Molekylformel C7 H5 N O
Molekylvikt (g/mol) 119.12
IUPAC-namn phenyl cyanate
LEDER N#COc1ccccc1
Specifikationer
Kemiskt namn eller material Phenyl Cyanate
Kvantitet 5 g
InChI formel InChI=1S/C7H5NO/c8-6-9-7-4-2-1-3-5-7/h1-5H
Formel vikt 119.0371