missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phenyl methacrylate, 95%, stab.

Catalog Number p-7045784
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2177-70-0
C10H10O2
162.188
MFCD00048117
QIWKUEJZZCOPFV-UHFFFAOYSA-N
phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
75121
phenyl 2-methylprop-2-enoate
CC(=C)C(=O)OC1=CC=CC=C1
This item is not returnable. View return policy
2177-70-0
C10H10O2
162.188
MFCD00048117
QIWKUEJZZCOPFV-UHFFFAOYSA-N
phenyl methacrylate, phenylmethacrylate, methacrylic acid, phenyl ester, 2-propenoic acid, 2-methyl-, phenyl ester, acmc-20ajx1, phenyl 2-methylacrylate #, methacrylic acid phenyl ester, phenyl methacrylate stabilized with bht
75121
phenyl 2-methylprop-2-enoate
CC(=C)C(=O)OC1=CC=CC=C1
Applications
Phenyl methacrylate is used as thermoset acrylic resin, fiber finishing agent and comonomers of resin lens.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY
Density 1.052
Boiling Point 115°C to 116°C (10 mmHg)
Flash Point 60°C (140°F)
Refractive Index 1.515
Quantity 5 g
UN Number UN3272
Beilstein 1907503
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material Phenyl methacrylate, stabilized
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'