Läs mer
Phenylacetyl CoA, TRC
Produktkod. 30418375
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30418375 10 mg
1 options
Produktkod. 30418375 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31920510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phenylacetyl CoA
CAS 7532-39-0
Molekylformel C29 H42 N7 O17 P3 S
Kvantitet 10 mg
Synonym Coenzyme A S-(Phenylacetate), Phenylthio-acetic Acid S-Ester With Coenzyme A, Phenylacetyl Coenzyme A, S-(Benzeneacetate) Coenzyme A
InChI formel InChI=1S/C29H42N7O17P3S/c1-29(2,24(40)27(41)32-9-8-19(37)31-10-11-57-20(38)12-17-6-4-3-5-7-17)14-50-56(47,48)53-55(45,46)49-13-18-23(52-54(42,43)44)22(39)28(51-18)36-16-35-21-25(30)33-15-34-26(21)36/h3-7,15-16,18,22-24,28,39-40H,8-14H2,1-2H3,(H,31,37)(H,32,41)(H,45,46)(H,47,48)(H2,30,33,34)(H2,42,43,44)/t18-,22-,23-,24+,28-/m1/s1
LEDER CC(C)(COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23)[C@@H](O)C(=O)NCCC(=O)NCCSC(=O)Cc4ccccc4
IUPAC-namn S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] 2-phenylethanethioate
Molekylvikt (g/mol) 885.667
Formel vikt 885.157